Assay Details
| Gene Symbol: THAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003432679.4 | XP_003432727.1 | 2 | 452 | 104 | |
| XM_005616676.3 | XP_005616733.1 | 3 | 545 | 104 | ||
| XM_022404349.1 | XP_022260057.1 | 3 | 605 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022404349.1 transcripts.
Set Membership: |
|

THAP8
Genomic Map