Assay Details
| Gene Symbol: KBTBD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014106408.2 | XP_013961883.1 | 1 | 1960 | 155 | |
| XM_014106409.2 | XP_013961884.1 | 1 | 1960 | 155 | ||
More Information
Set Membership: |
|

KBTBD6
Genomic Map