Assay Details
| Gene Symbol: TTLL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022418882.1 | XP_022274590.1 | 1056 | 140 | ||
| XM_022418883.1 | XP_022274591.1 | 1060 | 140 | |||
| XM_022418884.1 | XP_022274592.1 | 1054 | 140 | |||
| XM_022418885.1 | XP_022274593.1 | 2976 | 140 | |||
| XM_022418886.1 | XP_022274594.1 | 1059 | 140 | |||
| XM_022418887.1 | XP_022274595.1 | 1055 | 140 | |||
| XM_022418888.1 | XP_022274596.1 | 2635 | 140 | |||
| XM_022418889.1 | XP_022274597.1 | 457 | 140 | |||
More Information
Set Membership: |
|

TTLL10
Genomic Map