Assay Details
| Gene Symbol: KLHDC8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014122217.2 | XP_013977692.1 | 4 | 1205 | 100 | |
| XM_022406739.1 | XP_022262447.1 | 4 | 1222 | 100 | ||
| XM_022406740.1 | XP_022262448.1 | 3 | 1055 | 100 | ||
| XM_541887.6 | XP_541887.2 | 4 | 1218 | 100 | ||
More Information
Set Membership: |
|

KLHDC8B
Genomic Map