Assay Details
| Gene Symbol: ITPRIPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630387.3 | XP_005630444.1 | 2 | 1699 | 107 | |
| XM_005630388.3 | XP_005630445.1 | 2 | 1702 | 107 | ||
| XM_014120517.2 | XP_013975992.1 | 2 | 1601 | 107 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ITPRIPL1
Genomic Map