Assay Details
| Gene Symbol: TSPAN33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628407.3 | XP_005628464.1 | 1 - 2 | 270 | 125 | |
| XM_532430.6 | XP_532430.2 | 1 - 2 | 267 | 128 | ||
More Information
Set Membership: |
|

TSPAN33
Genomic Map