Assay Details
| Gene Symbol: MYLK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022427875.1 | XP_022283583.1 | 8 - 9 | 2044 | 140 | |
| XM_532569.6 | XP_532569.4 | 8 - 9 | 2026 | 140 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MYLK3
Genomic Map