Assay Details
| Gene Symbol: ZNF438 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617040.3 | XP_005617097.1 | 294 | 174 | ||
| XM_005617041.3 | XP_005617098.1 | 528 | 174 | |||
| XM_005617042.3 | XP_005617099.1 | 572 | 174 | |||
| XM_005617043.3 | XP_005617100.1 | 808 | 174 | |||
| XM_005617044.2 | XP_005617101.1 | 442 | 174 | |||
| XM_022406991.1 | XP_022262699.1 | 561 | 174 | |||
| XM_022406994.1 | XP_022262702.1 | 588 | 174 | |||
| XM_022406997.1 | XP_022262705.1 | 582 | 174 | |||
| XM_022406999.1 | XP_022262707.1 | 346 | 174 | |||
| XM_022407012.1 | XP_022262720.1 | 438 | 174 | |||
| XM_022407018.1 | XP_022262726.1 | 524 | 174 | |||
More Information
Set Membership: |
|

ZNF438
Genomic Map