Assay Details
| Gene Symbol: VSX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014116059.2 | XP_013971534.1 | 1 | 447 | 96 | |
| XM_547902.6 | XP_547902.3 | 1 | 447 | 96 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: VSX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014116059.2 | XP_013971534.1 | 1 | 447 | 96 | |
| XM_547902.6 | XP_547902.3 | 1 | 447 | 96 | ||
Set Membership: |
|