Assay Details
| Gene Symbol: ABHD15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022423393.1 | XP_022279101.1 | 2 | 1186 | 79 | |
| XM_548299.6 | XP_548299.2 | 2 | 1186 | 79 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ABHD15
Genomic Map