Assay Details
| Gene Symbol: RINL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616608.3 | XP_005616665.1 | 9 | 1539 | 153 | |
| XM_541634.5 | XP_541634.3 | 9 | 1394 | 153 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RINL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616608.3 | XP_005616665.1 | 9 | 1539 | 153 | |
| XM_541634.5 | XP_541634.3 | 9 | 1394 | 153 | ||
Set Membership: |
|