Assay Details
| Gene Symbol: SPATA21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617956.3 | XP_005618013.1 | 11 | 1950 | 167 | |
| XM_005617957.3 | XP_005618014.1 | 11 | 1947 | 167 | ||
| XM_022414215.1 | XP_022269923.1 | 11 | 1947 | 167 | ||
| XM_022414219.1 | XP_022269927.1 | 12 | 1787 | 167 | ||
| XM_022414221.1 | XP_022269929.1 | 11 | 1952 | 167 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SPATA21
Genomic Map