Assay Details
| Gene Symbol: GABRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619234.3 | XP_005619291.1 | 9 | 1457 | 236 | |
| XM_546259.6 | XP_546259.1 | 9 | 1573 | 212 | ||
| XM_861944.5 | XP_867037.1 | 10 | 1600 | 236 | ||
More Information
Set Membership: |
|

GABRG2
Genomic Map