Assay Details
| Gene Symbol: COLEC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022404410.1 | XP_022260118.1 | 833 | 153 | ||
| XM_022404411.1 | XP_022260119.1 | 797 | 153 | |||
| XM_022404412.1 | XP_022260120.1 | 805 | 153 | |||
| XM_022404413.1 | XP_022260121.1 | 802 | 153 | |||
| XM_022404414.1 | XP_022260122.1 | 768 | 153 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

COLEC11
Genomic Map