Assay Details
| Gene Symbol: ZSWIM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616367.3 | XP_005616424.1 | 3 - 4 | 970 | 75 | |
| XM_005616368.3 | XP_005616425.1 | 3 - 4 | 770 | 75 | ||
| XM_005616369.3 | XP_005616426.1 | 3 - 4 | 774 | 75 | ||
| XM_014119230.2 | XP_013974705.1 | 3 - 4 | 1000 | 75 | ||
| XM_014119234.2 | XP_013974709.1 | 3 - 4 | 706 | 75 | ||
More Information
Set Membership: |
|

ZSWIM9
Genomic Map