Assay Details
| Gene Symbol: HSPE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014111037.2 | XP_013966512.1 | 8 - 9 | 776 | 149 | |
| XM_536018.6 | XP_536018.2 | 7 - 8 | 638 | 149 | ||
| XM_852308.5 | XP_857401.2 | 6 - 7 | 581 | 149 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HSPE1
Genomic Map