Assay Details
| Gene Symbol: SLC16A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637517.2 | XP_005637574.1 | 5 | 685 | 141 | |
| XM_005637518.2 | XP_005637575.1 | 6 | 969 | 141 | ||
| XM_005637520.2 | XP_005637577.1 | 5 | 969 | 141 | ||
| XM_005637521.2 | XP_005637578.1 | 5 | 937 | 141 | ||
| XM_005637522.3 | XP_005637579.1 | 5 | 685 | 141 | ||
| XM_005637523.3 | XP_005637580.1 | 5 | 886 | 141 | ||
| XM_005637524.3 | XP_005637581.1 | 5 | 755 | 141 | ||
| XM_005637525.2 | XP_005637582.1 | 4 | 541 | 141 | ||
| XM_005637526.2 | XP_005637583.1 | 5 | 685 | 141 | ||
More Information
Set Membership: |
|

SLC16A12
Genomic Map