Assay Details
| Gene Symbol: NR3C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302828.1 | NP_001289757.1 | 1058 | 108 | ||
| XM_005617335.3 | XP_005617392.1 | 1590 | 108 | |||
| XM_014106563.2 | XP_013962038.1 | 1198 | 108 | |||
| XM_014106564.2 | XP_013962039.1 | 1343 | 108 | |||
| XM_014106568.2 | XP_013962043.1 | 1198 | 108 | |||
| XM_022405505.1 | XP_022261213.1 | 1189 | 108 | |||
| XM_535225.5 | XP_535225.2 | 1957 | 108 | |||
| GenBank mRNA | AB874470.1 | - | 1058 | 108 | ||
| KM496461.1 | - | 1957 | 108 | |||
| KM496462.1 | - | 1172 | 108 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NR3C1
Genomic Map