Assay Details
| Gene Symbol: KLF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022406293.1 | XP_022262001.1 | 1127 | 135 | ||
| XM_022406294.1 | XP_022262002.1 | 1078 | 135 | |||
| XM_846268.4 | XP_851361.1 | 1455 | 135 | |||
More Information
Set Membership: |
|

KLF15
Genomic Map