Assay Details
| Gene Symbol: MKRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005632138.3 | XP_005632195.1 | 979 | 154 | ||
| XM_014121597.2 | XP_013977072.1 | 1188 | 154 | |||
| XM_014121598.2 | XP_013977073.1 | 818 | 154 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MKRN2
Genomic Map