Assay Details
Gene Symbol: GHITM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619100.3 | XP_005619157.1 | 6 - 7 | 776 | 134 | |
XM_005619101.3 | XP_005619158.1 | 6 - 7 | 782 | 134 | ||
XM_005619102.3 | XP_005619159.1 | 6 - 7 | 652 | 134 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
