Assay Details
| Gene Symbol: CSDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022424371.1 | XP_022280079.1 | 2 - 3 | 432 | 73 | |
| XM_538352.6 | XP_538352.4 | 2 - 3 | 430 | 73 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CSDC2
Genomic Map