Assay Details
| Gene Symbol: RAB30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633430.3 | XP_005633487.1 | 4 - 5 | 616 | 132 | |
| XM_005633431.3 | XP_005633488.1 | 4 - 5 | 555 | 132 | ||
| XM_022407587.1 | XP_022263295.1 | 4 - 5 | 514 | 132 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RAB30
Genomic Map