Assay Details
| Gene Symbol: MAGED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641394.3 | XP_005641451.1 | 11 - 12 | 1487 | 167 | |
| XM_005641395.3 | XP_005641452.1 | 11 - 12 | 1543 | 167 | ||
| XM_846856.5 | XP_851949.1 | 11 - 12 | 1569 | 167 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MAGED2
Genomic Map