Assay Details
| Gene Symbol: CPEB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022411917.1 | XP_022267625.1 | 3 - 4 | 1350 | 105 | |
| XM_022411918.1 | XP_022267626.1 | 3 - 4 | 1351 | 105 | ||
| XM_022411919.1 | XP_022267627.1 | 3 - 4 | 1350 | 105 | ||
| XM_022411921.1 | XP_022267629.1 | 3 - 4 | 1349 | 105 | ||
| XM_022411922.1 | XP_022267630.1 | 3 - 4 | 1348 | 105 | ||
| XM_022411923.1 | XP_022267631.1 | 2 - 3 | 1205 | 105 | ||
| XM_022411924.1 | XP_022267632.1 | 2 - 3 | 1264 | 105 | ||
| XM_022411925.1 | XP_022267633.1 | 3 - 4 | 1291 | 105 | ||
| XM_022411926.1 | XP_022267634.1 | 2 - 3 | 1147 | 105 | ||
| XM_022411927.1 | XP_022267635.1 | 2 - 3 | 1138 | 105 | ||
More Information
Set Membership: |
|

CPEB3
Genomic Map