Assay Details
| Gene Symbol: RNF167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619921.3 | XP_005619978.1 | 3 - 4 | 303 | 154 | |
| XM_546574.5 | XP_546574.2 | 3 - 4 | 546 | 154 | ||
More Information
Set Membership: |
|

RNF167
Genomic Map