Assay Details
| Gene Symbol: KIF27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005615816.3 | XP_005615873.1 | 3 - 4 | 1101 | 121 | |
| XM_022420679.1 | XP_022276387.1 | 3 - 4 | 1130 | 121 | ||
| XM_022420685.1 | XP_022276393.1 | 3 - 4 | 766 | 121 | ||
More Information
Set Membership: |
|

KIF27
Genomic Map