Assay Details
| Gene Symbol: NSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629853.2 | XP_005629910.1 | 1592 | 67 | ||
| XM_005629854.2 | XP_005629911.1 | 1592 | 67 | |||
| XM_005629855.2 | XP_005629912.1 | 1592 | 67 | |||
| XM_005629856.2 | XP_005629913.1 | 1592 | 67 | |||
| XM_014120026.2 | XP_013975501.1 | 1592 | 67 | |||
| XM_852077.4 | XP_857170.1 | 1592 | 67 | |||
More Information
Set Membership: |
|

NSD3
Genomic Map