Assay Details
| Gene Symbol: RBFOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621565.3 | XP_005621622.1 | 8 - 9 | 884 | 109 | |
| XM_005621566.3 | XP_005621623.1 | 8 - 9 | 882 | 109 | ||
| XM_005621567.3 | XP_005621624.1 | 8 - 9 | 881 | 109 | ||
| XM_005621568.3 | XP_005621625.1 | 8 - 9 | 879 | 109 | ||
| XM_005621569.3 | XP_005621626.1 | 8 - 9 | 886 | 109 | ||
| XM_005621570.3 | XP_005621627.1 | 8 - 9 | 884 | 109 | ||
| XM_005621571.3 | XP_005621628.1 | 8 - 9 | 883 | 109 | ||
| XM_005621572.3 | XP_005621629.1 | 8 - 9 | 881 | 109 | ||
| XM_005621573.3 | XP_005621630.1 | 5 - 6 | 1253 | 109 | ||
| XM_005621574.3 | XP_005621631.1 | 2 - 3 | 738 | 109 | ||
| XM_005621575.3 | XP_005621632.1 | 2 - 3 | 653 | 109 | ||
| XM_014114639.2 | XP_013970114.1 | 8 - 9 | 883 | 109 | ||
| XM_846368.5 | XP_851461.1 | 2 - 3 | 635 | 109 | ||
More Information
Set Membership: |
|

RBFOX1
Genomic Map