Assay Details
| Gene Symbol: JPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638053.1 | XP_005638110.1 | 4 - 5 | 2198 | 107 | |
| XM_005638054.1 | XP_005638111.1 | 4 - 5 | 2198 | 107 | ||
| XM_022412295.1 | XP_022268003.1 | 4 - 5 | 2198 | 107 | ||
| XM_544130.4 | XP_544130.3 | 4 - 5 | 2198 | 107 | ||
| XM_844878.3 | XP_849971.1 | 4 - 5 | 2198 | 107 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

JPH1
Genomic Map