Assay Details
| Gene Symbol: DDX55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022410524.1 | XP_022266232.1 | 617 | 77 | ||
| XM_022410525.1 | XP_022266233.1 | 617 | 77 | |||
| XM_022410526.1 | XP_022266234.1 | 718 | 77 | |||
| XM_022410527.1 | XP_022266235.1 | 617 | 77 | |||
| XM_543371.5 | XP_543371.2 | 617 | 77 | |||
More Information
Set Membership: |
|

DDX55
Genomic Map