Assay Details
| Gene Symbol: NR2F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_536178.6 | XP_536178.2 | 2 - 3 | 1131 | 119 | |
| XM_844404.6 | XP_849497.2 | 2 - 3 | 1816 | 119 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NR2F2
Genomic Map