Assay Details
| Gene Symbol: CADM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014111257.1 | XP_013966732.1 | 8 - 9 | 1125 | 112 | |
| XM_022415312.1 | XP_022271020.1 | 8 - 9 | 1053 | 112 | ||
| XM_022415313.1 | XP_022271021.1 | 8 - 9 | 1132 | 112 | ||
| XM_022415314.1 | XP_022271022.1 | 8 - 9 | 1011 | 112 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CADM3
Genomic Map