Assay Details
| Gene Symbol: SLC26A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618676.3 | XP_005618733.1 | 3 - 4 | 2364 | 83 | |
| XM_545984.7 | XP_545984.3 | 2 - 3 | 2242 | 83 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC26A1
Genomic Map