Assay Details
| Gene Symbol: TENT4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617576.2 | XP_005617633.1 | 5 - 6 | 1025 | 137 | |
| XM_005617577.2 | XP_005617634.1 | 5 - 6 | 1034 | 137 | ||
| XM_022411501.1 | XP_022267209.1 | 5 - 6 | 1028 | 137 | ||
| XM_535307.6 | XP_535307.5 | 5 - 6 | 1028 | 137 | ||
More Information
Set Membership: |
|

TENT4B
Genomic Map