Assay Details
| Gene Symbol: MBOAT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003432706.4 | XP_003432754.1 | 5 - 6 | 710 | 65 | |
| XM_005616140.3 | XP_005616197.1 | 5 - 6 | 574 | 65 | ||
| XM_005616141.3 | XP_005616198.1 | 5 - 6 | 520 | 65 | ||
| XM_014118467.1 | XP_013973942.1 | 2 - 3 | 276 | 65 | ||
| XM_022427454.1 | XP_022283162.1 | 5 - 6 | 712 | 65 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MBOAT7
Genomic Map