Assay Details
| Gene Symbol: TMEM163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014121527.2 | XP_013977002.2 | 7 - 8 | 2848 | 102 | |
| XM_022406108.1 | XP_022261816.1 | 9 - 10 | 1399 | 102 | ||
| XM_022406109.1 | XP_022261817.1 | 7 - 8 | 894 | 102 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMEM163
Genomic Map