Assay Details
| Gene Symbol: UNC93B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631395.3 | XP_005631452.1 | 2 - 3 | 280 | 134 | |
| XM_540813.6 | XP_540813.3 | 1 - 2 | 371 | 134 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

UNC93B1
Genomic Map