Assay Details
| Gene Symbol: TTC25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624477.3 | XP_005624534.1 | 1290 | 83 | ||
| XM_005624478.3 | XP_005624535.1 | 1293 | 83 | |||
| XM_022422990.1 | XP_022278698.1 | 1161 | 83 | |||
More Information
Set Membership: |
|

TTC25
Genomic Map