Assay Details
| Gene Symbol: TMEM120A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620979.3 | XP_005621036.1 | 579 | 89 | ||
| XM_005620980.3 | XP_005621037.1 | 528 | 89 | |||
| XM_022419582.1 | XP_022275290.1 | 835 | 89 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMEM120A
Genomic Map