Assay Details
| Gene Symbol: IMMP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628736.3 | XP_005628793.1 | 3 - 4 | 229 | 116 | |
| XM_022427587.1 | XP_022283295.1 | 4 - 5 | 323 | 116 | ||
| XM_022427588.1 | XP_022283296.1 | 3 - 4 | 258 | 116 | ||
| XM_022427589.1 | XP_022283297.1 | 3 - 4 | 258 | 116 | ||
| XM_022427590.1 | XP_022283298.1 | 3 - 4 | 258 | 116 | ||
| XM_022427591.1 | XP_022283299.1 | 3 - 4 | 258 | 116 | ||
| XM_849634.3 | XP_854727.2 | 3 - 4 | 258 | 116 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IMMP2L
Genomic Map