Assay Details
| Gene Symbol: MID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003435641.4 | XP_003435689.1 | 5 - 6 | 1188 | 96 | |
| XM_005641647.3 | XP_005641704.1 | 5 - 6 | 1189 | 96 | ||
| XM_014111776.2 | XP_013967251.1 | 5 - 6 | 1151 | 96 | ||
| XM_014111777.2 | XP_013967252.1 | 5 - 6 | 1164 | 96 | ||
| XM_014111778.2 | XP_013967253.1 | 5 - 6 | 1406 | 96 | ||
| XM_014111779.2 | XP_013967254.1 | 5 - 6 | 1263 | 96 | ||
| XM_022416161.1 | XP_022271869.1 | 5 - 6 | 1685 | 96 | ||
| XM_022416162.1 | XP_022271870.1 | 4 - 5 | 1082 | 96 | ||
More Information
Set Membership: |
|

MID2
Genomic Map