Assay Details
| Gene Symbol: AZIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293659.1 | NP_001280588.1 | 5 - 6 | 769 | 129 | |
| XM_022405531.1 | XP_022261239.1 | 4 - 5 | 726 | 129 | ||
| XM_022405534.1 | XP_022261242.1 | 4 - 5 | 726 | 129 | ||
| XM_022405539.1 | XP_022261247.1 | 4 - 5 | 726 | 129 | ||
More Information
Set Membership: |
|

AZIN2
Genomic Map