Assay Details
| Gene Symbol: TWIST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635928.2 | XP_005635985.1 | 1 | 292 | 52 | |
| XM_543311.5 | XP_543311.1 | 1 | 290 | 52 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TWIST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635928.2 | XP_005635985.1 | 1 | 292 | 52 | |
| XM_543311.5 | XP_543311.1 | 1 | 290 | 52 | ||
Set Membership: |
|