Assay Details
| Gene Symbol: URI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616795.3 | XP_005616852.1 | 5 - 6 | 627 | 127 | |
| XM_014120925.1 | XP_013976400.1 | 4 - 5 | 285 | 127 | ||
| XM_022426483.1 | XP_022282191.1 | 5 - 6 | 675 | 127 | ||
More Information
Set Membership: |
|

URI1
Genomic Map