Assay Details
| Gene Symbol: PPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627173.2 | XP_005627230.1 | 7 - 8 | 858 | 103 | |
| XM_014118353.2 | XP_013973828.1 | 7 - 8 | 867 | 103 | ||
| XM_022425867.1 | XP_022281575.1 | 6 - 7 | 901 | 103 | ||
| XM_532090.6 | XP_532090.2 | 7 - 8 | 1095 | 103 | ||
More Information
Set Membership: |
|

PPT2
Genomic Map