Assay Details
| Gene Symbol: MTFMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109514.2 | XP_013964989.1 | 1 - 2 | 227 | 146 | |
| XM_848312.5 | XP_853405.3 | 1 - 2 | 227 | 146 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MTFMT
Genomic Map