Assay Details
| Gene Symbol: ORMDL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624545.3 | XP_005624602.1 | 433 | 121 | ||
| XM_005624546.3 | XP_005624603.1 | 332 | 121 | |||
| XM_005624547.3 | XP_005624604.1 | 428 | 121 | |||
| XM_846217.5 | XP_851310.1 | 351 | 121 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ORMDL3
Genomic Map