Assay Details
| Gene Symbol: RHEBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636880.3 | XP_005636937.1 | 1 - 2 | 288 | 147 | |
| XM_846022.4 | XP_851115.1 | 1 - 2 | 286 | 147 | ||
More Information
Set Membership: |
|

RHEBL1
Genomic Map