Assay Details
| Gene Symbol: TBC1D20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014107264.2 | XP_013962739.1 | 5 - 6 | 800 | 106 | |
| XM_856834.4 | XP_861927.2 | 4 - 5 | 666 | 106 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TBC1D20
Genomic Map